RuBP-4S fluoreszierender Farbstoff, für die Histologie

Lieferant: Biotium

Synonyme: [Ruthenium(bathophenanthroline)(bis(bathophenanthrolinedisulphonate))],disodium salt

80025
80025.EA 340 EUR
80025.
RuBP-4S fluoreszierender Farbstoff, für die Histologie
RuBP-4S

RuBP-4S ([Ruthenium(bathophenanthroline)(bis(bathophenanthrolinedisulfonate))],disodium salt) is a luminescent ruthenium complex similar to RuBPS (80021). Two of the bathophenanthroline ligands are bis-sulfonated while the third bathophenanthroline ligand is neutral, resulting in an overall double negative charges of the ruthenium complex.


  • λEx/λ Em(H2O)= 462 (broad)/603 nm
  • Yellow solid soluble in water
  • Store at 4°C and protect from light


RuBP-4S shares the spectral properties of RuBPS and may also have similar applications.

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9183bfa4aafbaea47b66f3e140039a5fffb8d047&vid=455a0b4ebd59e4569e7401578f67c69515f13df0&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR