Aceton, BAKER, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Propanon, DMK, 2-Propanon, Dimethylketon

8003.1000EA 39.7 EUR
8003.1000 8003.2500 8003.5000 8003.9025 8003.9010 8003.9200ST
Aceton, BAKER, J.T.Baker®
Aceton
Formel: CH₃COCH₃
Siedepunkt: 56,2 °C (1013 hPa)
Schmelzpunkt: –95,4 °C
Dichte: 0,792 g/cm³ (20 °C)
Flammpunkt: <–20 °C (geschlossener Tiegel)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008765
CAS-Nummer: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use
Acidity or Alkalinity passes test
Appearance of solution passes test
Identification passes test
Matter Insoluble in Water passes test
Reducing Substances passes test
Relative Density 0.790 - 0.793
Residue on Evaporation max. 50 ppm
Water (H2O) max. 3 g/l
Related Substances:
Benzene max. 2 ppm (v/v)
Methanol (CH3OH) max. 0.05 % (v/v)
Other Impurities passes test
2-Propanol (CH3CHOHCH3) max. 0.05 % (v/v)
Additional Specification(s):
Assay (by GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR