Biotin-CF® 640R fluoreszierender Farbstoff
Lieferant: Biotium
80032
80032.EA 336 EUR
80032.
Biotin-CF® 640R fluoreszierender Farbstoff
Biotin-CF®
Potential applications of Biotin-CF® dye include detection of biotin binding sites and the degree of biotinylation of proteins, and for the measurement of avidin and streptavidin in crude biological samples. In addition, biotin-CF® dye can be used as a polar tracer by microinjection to study the morphology of cells. CF® dyes are very photostable which make them suitable for studies where prolonged exposure to light may be necessary.
  • Ex/Em 642/662 nm
  • Dark Blue solid soluble in DMSO or DMF
CF® Dye Biotin is available with the following CF® dyes: CF®568, CF®633, and CF®640R.
Bitte beachten Store desiccated at 4 °C. Protect from light.