Sulphorhodamine 101 fluoreszierender Farbstoff, für die Histologie
Lieferant: Biotium
Synonyme:
Sulforhodamine 101, Texas Red, SR 101, Sulforhodamine 640, 9-(2,4-Disulphophenyl)-2,3,6,7,12,13,16,17-octahydro-1H,5H,11H,15H-xantheno[2,3,4-ij:5,6,7-i'j']diquinolizin-18-ium inner salt
80101
80101.EA
336
EUR
80101.
Sulphorhodamine 101 fluoreszierender Farbstoff, für die Histologie
Sulphorhodamine 101
Sulforhodamine 101 has a longer absorption wavelength than does sulforhodamine B. Like sulforhodamine B and G, in addition to their potential use in cancer drug screening, these fluorescent dyes have been primarily used as polar tracers for the studies of neuronal cell morphology and cell-cell communication.
- λEx/λEm = 586/605 nm
- Red solid soluble in water
- Store at room temperature
Formel:
C₃₁H₃₀N₂O₇S₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012407 CAS-Nummer: 60311-02-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...