Sulphorhodamine G fluoreszierender Farbstoff, für die Histologie
Lieferant: Biotium
80102
80102.EA
208
EUR
80102.
Sulphorhodamine G fluoreszierender Farbstoff, für die Histologie
Sulphorhodamine G
Similar to other sulforhodamine dyes, the highly water-soluble sulforhodamine G can also be used as a polar tracer for the studies of cell morphology and neuronal cell-cell communication.
- λEx/λ Em = 529/548 nm
- Orange red solid soluble in water
- Store at room temperature
Lagertemperatur:
Raumtemperatur |
MDL:
MFCD00213704 CAS-Nummer: 5873-16-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...