N-tert-Butyl-α-Phenylnitron ≥99,5% (durch HPLC) für ESR-Spektroskopie, Supelco®
Lieferant: Merck
Synonyme:
PBN, N-Benzylidene-tert-butylamine N-Oxide, N-tert-Butyl-α-phenylnitrone
80126-1G
80126-1GEA
181
EUR
80126-1G
N-tert-Butyl-α-Phenylnitron ≥99,5% (durch HPLC) für ESR-Spektroskopie, Supelco®
N-tert-Butyl-α-Phenylnitron
N-tert-Butyl-a-phenylnitrone was used as spin trapping agent during measurement of scavenging rate constant of carotenoid using EPR spin-trapping technique.
his reagent helps in obtaining a six-line ESR spectrum and hyperfine coupling constants, confirming the presence of carbon-based radical in uric acid and peroxynitrite, using electron spin resonance spectroscopy and liquid chromatography mass spectrometry
Formel:
C₁₁H₁₅NO Lagertemperatur: Gefrierschrank |
MDL:
MFCD00008799 CAS-Nummer: 3376-24-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...