Butylacetat, BAKER, Laboratory Reagent, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
Synonyme:
Essigsäurebutylester
8018.1000EA
75.27
EUR
8018.1000
Butylacetat, BAKER, Laboratory Reagent, J.T.Baker®
Butylacetat
Formel:
CH₃COO(CH₂)₃CH₃ Molecular Weight: 116.16 g/mol Siedepunkt: 126 Schmelzpunkt: –76 °C Dichte: 0.875 g/cm³ (20 °C) Flammpunkt: 25 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009445 CAS-Nummer: 123-86-4 EINECS: 204-658-1 UN: 1123 ADR: 3 III Merck Index: 13,01534 |
|
Spezifikation Testergebnisse
Assay | min. 98 % |
Acidity (as CH3COOH) | max. 0.005 % |
Boiling Range | 124 - 127 °C |
Density (g/ml) at 20°C | 0.878 - 0.881 |
n-Butanol | max. 0.5 % |
Residue after Evaporation | max. 0.005 % |
Water (H2O) | max. 0.05 % |
Trace Impurities (in ppm) | |
Iron (Fe) | max. 0.5 |
Heavy Metals (as Pb) | max. 0.5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...