Cyclohexan, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Gefahr

8026.1000EA 79.78 EUR
8026.1000 8026.2500 8026.9025
Cyclohexan, BAKER ANALYZED® ACS, J.T.Baker®
Cyclohexan
Formel: C₆H₁₂
Siedepunkt: 81 °C (1013 hPa)
Schmelzpunkt: 7 °C
Dichte: 0,7785 g/cm³ (20 °C)
Flammpunkt: –18 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003814
CAS-Nummer: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
Merck Index: 13,02752
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a1e51ba998bef126804c3fd25b361be72cd9545&vid=d1e7f75d9f2a944eb15a7740c5254588cc9a4e67&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Exceeds ACS Specifications
Assay (by GC) min. 99.0 %
Benzene (C6H6) max. 0.005 %
Color (APHA) max. 10
Residue after Evaporation max. 0.002 %
Substances Darkened by H2SO4 passes test
Water (H2O) max. 0.02 %
Trace Impurities (in ppm)
Aluminium (Al) max. 0.5
Boron (B) max. 0.02
Barium (Ba) max. 0.1
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.05
Cobalt (Co) max. 0.02
Chromium (Cr) max. 0.02
Copper (Cu) max. 0.02
Iron (Fe) max. 0.1
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.02
Nickel (Ni) max. 0.02
Lead (Pb) max. 0.1
Tin (Sn) max. 0.1
Zinc (Zn) max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR