Natriumsulfat, wasserfrei ≥99,0% (Trockenmasse) USP, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials


Veränderungsmanagement-Kategorie= R

8028-06EA 173.31 EUR
8028-06 8028-88 8028-12 8028-65 8028-F9 MACR8028-20
Natriumsulfat, wasserfrei ≥99,0% (Trockenmasse) USP, Macron Fine Chemicals™

No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.


Metallic Residues: No metal catalysts or metal reagents, as defined by EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of this material.

Formel: Na₂SO₄
Siedepunkt: 1429 °C (1013 hPa)
Schmelzpunkt: 888 °C
Dichte: 2,7 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003504
CAS-Nummer: 7757-82-6
EINECS: 231-820-9
Merck Index: 12,08829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR