Supelco® TraceCERT® Organic Reference Standard, 2-Methylthiophene
Lieferant: Merck
80364-100MG
80364-100MGEA
119
EUR
80364-100MG
Supelco® TraceCERT® Organic Reference Standard, 2-Methylthiophene
Standards
Organische Standards
- GHS signal: GHS02
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Bitte beachten P210 - P370 + P378 - P403 + P235