n-Hexan, BAKER ANALYZED®, Laborreagens, J.T.Baker®

Lieferant: Avantor
Gefahr

8044.1000EA 76.1 EUR
8044.1000 8044.2500 8044.9025 8044.5000 8044.9200
n-Hexan, BAKER ANALYZED®, Laborreagens, J.T.Baker®
n-Hexan
Formel: H₃C(CH₂)₄CH₃
Siedepunkt: 69 °C (1013 hPa)
Dichte: 0,659 g/cm³ (20 °C)
Flammpunkt: –22 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009520
CAS-Nummer: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (by GC) min. 95 %
Aromatic Compounds (as C6H6) max. 0.02 %
Color (APHA) max. 10
Density (g/ml) at 25°C max. 0.657
Residue after Evaporation max. 0.001 %
Sulfur Compounds (as S) max. 0.005 %
Thiophene passes test
Water Soluble Titrable Acid (meq/g) max. 0.0003
Trace Impurities (in ppm)
Aluminium (Al) max. 0.5
Boron (B) max. 0.02
Barium (Ba) max. 0.1
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.05
Cobalt (Co) max. 0.02
Chromium (Cr) max. 0.02
Copper (Cu) max. 0.02
Iron (Fe) max. 0.1
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.02
Nickel (Ni) max. 0.02
Lead (Pb) max. 0.1
Tin (Sn) max. 0.1
Zinc (Zn) max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR