Kieselgel, spherical 40-75 μm particle size, Supelco®

Lieferant: Merck

53698-1KG 53698-5KG 80442-100G 80442-1KG
53698-1KGEA 423 EUR
53698-1KG 53698-5KG 80442-100G 80442-1KG
Kieselgel, spherical 40-75 μm particle size, Supelco®
Silica gel
Formel: SiO₂
Molecular Weight: 60.08 g/mol
Siedepunkt: 2230 °C
Schmelzpunkt: > 1600 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011232
CAS-Nummer: 63231-67-4
EINECS: 231-545-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f07db57db7fff80e643aea2a8687eceff7c31c45&vid=2c0ba06e92d734c5d6bb9cc529323e2cb1bef7b5&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9f215b0da26c79111e843dc89f7c0e84185a415&vid=8c01453499a59f4060ae9f1b8911f90fe474b578&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR