Natriumborhydrid 98,5-101,5% (gasvolumetrisch), LiChropur™ derivatisation reagent, for HPLC, Supelco®

Lieferant: Merck

80546-100G 80546-25G
80546-100GEA 373 EUR
80546-100G 80546-25G
Natriumborhydrid 98,5-101,5% (gasvolumetrisch), LiChropur™ derivatisation reagent, for HPLC, Supelco®
Natriumtetrahydroborat
Formel: NaBH₄
MDL: MFCD00003518
CAS-Nummer: 16940-66-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR