Organic reference standard, (-)-beta-Pinene, analytical standard, Supelco®
Lieferant: Merck
80609-1ML
80609-1MLEA 65 EUR
80609-1ML
Organic reference standard, (-)-beta-Pinene, analytical standard, Supelco®
Standards Organische Standards
(-)-β-Pinene is a monoterpenoid, naturally found in the resins of pine trees and other conifers. It comprises of an exocyclic double bond in its structure.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c29e25fc234c1597d3c54f7c205cf6d43ee3c8d6&vid=3e00887ec1263559f88a42bb17e84cc16e5e14e7&prid=web&clid=SAPDAM