1-Propanol ≥99,0%, BAKER ANALYZED®, Laborreagens, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: 1-Propylalkohol, 1-Hydroxypropan, Ethylcarbinol

8066.1000EA 88.72 EUR
8066.1000 8066.2500
1-Propanol ≥99,0%, BAKER ANALYZED®, Laborreagens, J.T.Baker®
1-Propanol
Formel: H₃CCH₂CH₂OH
Molecular Weight: 60.1 g/mol
Siedepunkt: 97
Schmelzpunkt: –127 °C
Dichte: 0.8053 g/cm³ (20 °C)
Flammpunkt: 15 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002941
CAS-Nummer: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3 II
Merck Index: 13,07934
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=deb28b985024604344065eea93ff6c6a9dd5a431&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (by GC) min. 99 %
Boiling Range max. 2.0 °C
Color (APHA) max. 10
Density (g/ml) at 25°C 0.800 - 0.804
Ethanol (C2H5OH) (by GC) max. 0.05 %
Free Acid (as C2H5COOH) max. 0.001 %
Methanol (by GC) (CH3OH) max. 0.05 %
Recorded Boiling Point 97.2 °C
Residue after Evaporation max. 0.005 %
Substances Darkened by H2SO4 passes test
Substances Reducing KMnO4 (as O) max. 0.0003 %
Water (H2O) max. 0.1 %
2-Propanol (C3H7OH) (by GC) max. 0.05 %
Trace Impurities (in ppm)
Aluminium (Al) max. 0.5
Boron (B) max. 0.02
Barium (Ba) max. 0.1
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.05
Cobalt (Co) max. 0.02
Chromium (Cr) max. 0.02
Copper (Cu) max. 0.02
Iron (Fe) max. 0.1
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.02
Nickel (Ni) max. 0.02
Lead (Pb) max. 0.1
Tin (Sn) max. 0.1
Zinc (Zn) max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR