Trifluoressigsäureanhydrid Acylierungsreagenz, zur GC-Derivatisierung

Lieferant: MACHEREY-NAGEL
Gefahr

Synonyme: Trifluoracetic anhydride, TFAA

701130.510 701130.110
807-0113EA 145 EUR
807-0113 MANA701130.110
Trifluoressigsäureanhydrid Acylierungsreagenz, zur GC-Derivatisierung
Trifluoressigsäureanhydrid

Acylation with fluorinated acid anhydrides can be used for alcohols, phenols, carboxylic acids, amines, amino acids and steroids forming volatile, stable derivatives suited for FID as well as for ECD detection.


  • Addition of halogenated carbons enhances detectability by GC-ECD
  • Derivatives are hydrolytical stable
  • Increased sensitivity by adding molecular weight

Formel: C₄F₆O₃
Molecular Weight: 210,03 g/mol
Siedepunkt: 39,5–40.5 °C (760 mm Hg)
Dichte: 1,490 g/cm³ (d20°/4°)
MDL: MFCD00000416
CAS-Nummer: 407-25-0
UN: 3316
ADR: 9,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR