Tetrahydrofuran stabilisiert, BAKER ANALYZED®, Laborreagens, J.T.Baker®

Lieferant: Avantor
Gefahr

8075.1000EA 106.34 EUR
8075.1000 8075.2500 8075.9025
Tetrahydrofuran stabilisiert, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Tetrahydrofuran
Formel: C₄H₈O
Siedepunkt: 66 °C (1013 hPa)
Schmelzpunkt: –108,5 °C
Dichte: 0,888 g/cm³ (20 °C)
Flammpunkt: –21,5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005356
CAS-Nummer: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (by GC) min. 99 %
Boiling Range 65 - 67 °C
Color (APHA) passes test
Density (g/ml) at 25°C 0.880 - 0.883
Peroxide (as H2O2) max. 0.02 %
Residue after Evaporation max. 0.002 %
Water (H2O) max. 0.02 %
Trace Impurities (in ppm)
Aluminium (Al) max. 0.5
Boron (B) max. 0.02
Barium (Ba) max. 0.1
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.05
Cobalt (Co) max. 0.02
Chromium (Cr) max. 0.02
Copper (Cu) max. 0.02
Iron (Fe) max. 0.1
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.02
Nickel (Ni) max. 0.02
Lead (Pb) max. 0.1
Tin (Sn) max. 0.1
Zinc (Zn) max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR