Xylol (Isomerengemisch), BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Gefahr

Synonyme: Dimethylbenzol

8080.1000EA 78.43 EUR
8080.1000 8080.2500 8080.9025 8080.5000
Xylol (Isomerengemisch), BAKER ANALYZED® ACS, J.T.Baker®
Xylol (Isomerengemisch)
Formel: C₆H₄(CH₃)₂
Molecular Weight: 106.17 g/mol
Siedepunkt: 140
Schmelzpunkt: –34 °C
Dichte: 0.86…0.88 g/cm³ (20 °C)
Flammpunkt: 25
Lagertemperatur: Raumtemperatur
MDL: MFCD00077264
CAS-Nummer: 1330-20-7
EINECS: 215-535-7
UN: 1307
ADR: 3 III
Merck Index: 13,10136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Exceeds ACS Specifications
Assay min. 98.5 %
Color (APHA) max. 10
Ethylbenzene max. 25 %
Residue after Evaporation max. 0.002 %
Sulfur Compounds (as S) max. 0.003 %
Substances Darkened by H2SO4 passes test
Water (H2O) max. 0.03 %
Trace Impurities (in ppm)
Aluminium (Al) max. 0.5
Boron (B) max. 0.02
Barium (Ba) max. 0.1
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.05
Cobalt (Co) max. 0.02
Chromium (Cr) max. 0.02
Copper (Cu) max. 0.02
Iron (Fe) max. 0.1
Magnesium (Mg) max. 0.1
Manganese (Mn) max. 0.02
Nickel (Ni) max. 0.02
Lead (Pb) max. 0.1
Tin (Sn) max. 0.1
Zinc (Zn) max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR