Xylol (Isomerengemisch), BAKER ANALYZED® ACS, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
8080.1000EA
78.43
EUR
8080.1000
8080.2500
8080.9025
8080.5000
Xylol (Isomerengemisch), BAKER ANALYZED® ACS, J.T.Baker®
Xylol (Isomerengemisch)
Formel:
C₆H₄(CH₃)₂ Siedepunkt: 140 °C (1013 hPa) Schmelzpunkt: –34 °C Dichte: 0,86…0.88 g/cm³ (20 °C) Flammpunkt: 25 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00077264 CAS-Nummer: 1330-20-7 EINECS: 215-535-7 UN: 1307 ADR: 3,III Merck Index: 13,10136 |
|
Spezifikation Testergebnisse
Exceeds ACS Specifications | |
Assay | min. 98.5 % |
Color (APHA) | max. 10 |
Ethylbenzene | max. 25 % |
Residue after Evaporation | max. 0.002 % |
Sulfur Compounds (as S) | max. 0.003 % |
Substances Darkened by H2SO4 | passes test |
Water (H2O) | max. 0.03 % |
Trace Impurities (in ppm) | |
Aluminium (Al) | max. 0.5 |
Boron (B) | max. 0.02 |
Barium (Ba) | max. 0.1 |
Calcium (Ca) | max. 0.5 |
Cadmium (Cd) | max. 0.05 |
Cobalt (Co) | max. 0.02 |
Chromium (Cr) | max. 0.02 |
Copper (Cu) | max. 0.02 |
Iron (Fe) | max. 0.1 |
Magnesium (Mg) | max. 0.1 |
Manganese (Mn) | max. 0.02 |
Nickel (Ni) | max. 0.02 |
Lead (Pb) | max. 0.1 |
Tin (Sn) | max. 0.1 |
Zinc (Zn) | max. 0.1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...