Supelco® TraceCERT® Organic Reference Standard, Pyridoxine hydrochloride
Lieferant: Merck
80823-50MG
80823-50MGEA 85 EUR
80823-50MG
Supelco® TraceCERT® Organic Reference Standard, Pyridoxine hydrochloride
Standards Organische Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Bitte beachten P280 - P305 + P351 + P338
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f485c0489a3ad8bd933ced3c49d8a0897a71bca5&vid=f485c0489a3ad8bd933ced3c49d8a0897a71bca5&prid=web&clid=SAPDAM