18-Krone-6 (1,4,7,10,13,16-Hexaoxacyclooctadecan) ≥99,0% (durch GC) für die Ionenchromatographie, Supelco®

Lieferant: Merck
Achtung

80833-10G
80833-10GEA 93.7 EUR
80833-10G
18-Krone-6 (1,4,7,10,13,16-Hexaoxacyclooctadecan) ≥99,0% (durch GC) für die Ionenchromatographie, Supelco®
18-Krone-6 (1,4,7,10,13,16-Hexaoxacyclooctadecan)

18-Crown-6 can be useful as phase-transfer catalysts.

Formel: C₁₂H₂₄O₆
Schmelzpunkt: 42,..45 °C (Lit.)
MDL: MFCD00005113
CAS-Nummer: 17455-13-9
EINECS: 241-473-5

Bestellen

Spezifikation Testergebnisse

Appearance (Color) White to Faint Yellow
Appearance (form) Powder or Solid or Crystals or Crystalline Chunk(s) or Chunk(s)
Purity (GC area %) ≥ 99.0 %
Carbon Content 54.0 - 55.1 %
Infrared Spectrum Conforms to Structure
Metal Trace Analysis (ICP) Corresponds to Requirements
Aluminium (ICP) ≤ 1 mg/kg
Barium (ICP) ≤ 1 mg/kg
Bismuth (ICP) ≤ 1 mg/kg
Calcium (ICP) ≤ 5 mg/kg
Cadmium (ICP) ≤ 1 mg/kg
Cobalt (ICP) ≤ 1 mg/kg
Chromium (ICP) ≤ 1 mg/kg
Copper (ICP) ≤ 1 mg/kg
Iron (ICP) ≤ 1 mg/kg
Potassium (ICP) ≤ 20 mg/kg
Lithium (ICP) ≤ 1 mg/kg
Magnesium (ICP) ≤ 1 mg/kg
Manganese (ICP) ≤ 1 mg/kg
Molybdenum (ICP) ≤ 1 mg/kg
Sodium (ICP) ≤ 20 mg/kg
Nickel (ICP) ≤ 1 mg/kg
Lead (ICP) ≤ 1 mg/kg
Strontium (ICP) ≤ 1 mg/kg
Zinc (ICP) ≤ 1 mg/kg
Ion Chromatography (NH₄⁺) ≤ 10 mg/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR