Scriber pen, Glascribe®, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
H441500000
811-0103EA 94.1 EUR
811-0103
Scriber pen, Glascribe®, SP Bel-Art
Schreibstifte Stylusstifte und Schreiber
The Glascribe® Pen, with retractable tungsten carbide tip, lets you inscribe on glass, ceramics, and plastic.
  • With pocket clip, so it is always handy when needed