Permanent marker pen, Edding 400
Lieferant: THEODOR KIHLER BURO EINRICHTUN
00440001 0440002 04400004
811-3131EA 8.3 EUR
811-3131 811-3132 811-3134
Permanent marker pen, Edding 400
Schreibstifte Permanentmarker
Permananent marker for writing, colouring, labelling for materials such as metal, glass or plastic, low odour ink, round interchangeable tip. Line thickness of 1 mm.
  • Fast drying
  • Waterproof
  • Refillable
Bestellen
TECHNISCHE DATEN
Environmentally Preferable