Vanadyl(IV)acetylacetonat
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
CAS No.: 3153-26-2
Molecular Formula: C10H14O5V
Formula Weight: 265.16
Physical Form: Powder
Density: 1.4
Melting Point: 256-259°
Boiling Point: 140°/13mm
Solubility: Moderately soluble in acetone, ether and chloroform. Soluble in ethanol and benzene
UN No.: 3285
MDL No.: MFCD00000032
As a catalyst, as a synthesis intermediate, as a paint dryer, and as a pigment
Formel:
C₁₀H₁₄O₅V Siedepunkt: 140 °C (13 mmHg) Schmelzpunkt: 256…259 °C Dichte: 1,4 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000032 CAS-Nummer: 3153-26-2 EINECS: 221-590-8 UN: 3285 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...