n-Pentan, BAKER ANALYZED®, Laborreagens, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
8114.2500EA
258.89
EUR
8114.2500
8114.1000
8114.9025
8114.9200
n-Pentan, BAKER ANALYZED®, Laborreagens, J.T.Baker®
n-Pentan
Formel:
H₃C(CH₂)₃CH₃ Siedepunkt: 35…36 °C (1013 hPa) Schmelzpunkt: –130 °C Dichte: 0,63 g/cm³ (20 °C) Flammpunkt: –40 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009498 CAS-Nummer: 109-66-0 EINECS: 203-692-4 UN: 1265 ADR: 3,II REACH: 01-2119459286-30 Merck Index: 13,07193 |
|
Spezifikation Testergebnisse
Assay (by GC) | min. 99 % |
Boiling Range | 35 - 37 °C |
Density (g/ml) at 20°C | 0.624 - 0.626 |
Residue after Evaporation | max. 0.001 % |
Sulfur Compounds (as S) | max. 0.005 % |
Water (H2O) | max. 0.01 % |
Trace Impurities (in ppm)Aluminium (Al) | max. 0.5 |
Trace Impurities (in ppm)Boron (B) | max. 0.02 |
Trace Impurities (in ppm)Barium (Ba) | max. 0.1 |
Trace Impurities (in ppm)Calcium (Ca) | max. 0.5 |
Trace Impurities (in ppm)Cadmium (Cd) | max. 0.05 |
Trace Impurities (in ppm)Cobalt (Co) | max. 0.02 |
Trace Impurities (in ppm)Chromium (Cr) | max. 0.02 |
Trace Impurities (in ppm)Copper (Cu) | max. 0.02 |
Trace Impurities (in ppm)Iron (Fe) | max. 0.1 |
Trace Impurities (in ppm)Magnesium (Mg) | max. 0.1 |
Trace Impurities (in ppm)Manganese (Mn) | max. 0.02 |
Trace Impurities (in ppm)Nickel (Ni) | max. 0.02 |
Trace Impurities (in ppm)Lead (Pb) | max. 0.1 |
Trace Impurities (in ppm)Tin (Sn) | max. 0.1 |
Trace Impurities (in ppm)Zinc (Zn) | max. 0.1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...