n-Pentan, BAKER ANALYZED®, Laborreagens, J.T.Baker®

Lieferant: Avantor
Gefahr

8114.2500EA 258.89 EUR
8114.2500 8114.1000 8114.9025 8114.9200
n-Pentan, BAKER ANALYZED®, Laborreagens, J.T.Baker®
n-Pentan
Formel: H₃C(CH₂)₃CH₃
Siedepunkt: 35…36 °C (1013 hPa)
Schmelzpunkt: –130 °C
Dichte: 0,63 g/cm³ (20 °C)
Flammpunkt: –40 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009498
CAS-Nummer: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3,II
REACH: 01-2119459286-30
Merck Index: 13,07193
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ba2c9c81a4b1f44769627a00ba49b6cc8e63132&vid=f6831eb15ab1602d8c752468dc3ee31cc4ee61c0&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (by GC) min. 99 %
Boiling Range 35 - 37 °C
Density (g/ml) at 20°C 0.624 - 0.626
Residue after Evaporation max. 0.001 %
Sulfur Compounds (as S) max. 0.005 %
Water (H2O) max. 0.01 %
Trace Impurities (in ppm)Aluminium (Al) max. 0.5
Trace Impurities (in ppm)Boron (B) max. 0.02
Trace Impurities (in ppm)Barium (Ba) max. 0.1
Trace Impurities (in ppm)Calcium (Ca) max. 0.5
Trace Impurities (in ppm)Cadmium (Cd) max. 0.05
Trace Impurities (in ppm)Cobalt (Co) max. 0.02
Trace Impurities (in ppm)Chromium (Cr) max. 0.02
Trace Impurities (in ppm)Copper (Cu) max. 0.02
Trace Impurities (in ppm)Iron (Fe) max. 0.1
Trace Impurities (in ppm)Magnesium (Mg) max. 0.1
Trace Impurities (in ppm)Manganese (Mn) max. 0.02
Trace Impurities (in ppm)Nickel (Ni) max. 0.02
Trace Impurities (in ppm)Lead (Pb) max. 0.1
Trace Impurities (in ppm)Tin (Sn) max. 0.1
Trace Impurities (in ppm)Zinc (Zn) max. 0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR