Poly(dimethylsiloxane) analytical standard, for GPC
Lieferant: Merck
81153-100MG
81153-100MGEA 503 EUR
81153-100MG
Poly(dimethylsiloxane) analytical standard, for GPC
Standards Gaschromatographie-Standards
Poly(dimethylsiloxane) [PDMS] is a polymer. It is a stable silicone rubber. It has been used as ion selective membrane for ion-sensitive field-effect transistors, as spring material in accelerometers, elastomer on a tactile sensor and as flexible encapsulation material in decouple sensors.
Standards: Exact values can be taken from the accompanying certificate of analysis
Bitte beachten Storage temperature: 2 to 8 °C. Limited shelf life, expiry date on the label.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab072b6d4ff7e446fd5a90d9f4aff87f229428ba&vid=57503b3ff18f58290464f45a7355c491eeca6007&prid=web&clid=SAPDAM