Poly(ethylene glycol) analytical standards, for GPC, Supelco
Lieferant: Merck
81201-500MG
81201-500MGEA
797
EUR
81201-500MG
Poly(ethylene glycol) analytical standards, for GPC, Supelco
Standards
Gaschromatographie-Standards
Poly(ethylene glycol) is the most widely used non-ionic, hydrophilic polymer, which can find wide applications in biomedical industry such as in drug delivery systems.
Standards: Exact values can be taken from the accompanying certificate of analysis