Polyethylenglycol 12.000, Supelco®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
81296-1G
81296-1GEA
226
EUR
81296-1G
Polyethylenglycol 12.000, Supelco®
Polyethylenglycol 12.000
Formel:
H(OCH₂CH₂)nOH Lagertemperatur: Raumtemperatur |
MDL:
MFCD00081839 CAS-Nummer: 25322-68-3 EINECS: 500-038-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...