Poly(styrenesulfonic acid sodium salt) analytical standards for GPC
Lieferant: Merck
81604-250MG 81606-250MG 81607-250MG 81609-250MG 81610-250MG 81612-250MG 81614-250MG 81617-250MG 04406-250MG
81604-250MGEA 147 EUR
81604-250MG 81606-250MG 81607-250MG 81609-250MG 81610-250MG 81612-250MG 81614-250MG 81617-250MG 04406-250MG
Poly(styrenesulfonic acid sodium salt) analytical standards for GPC
Standards Gaschromatographie-Standards
Standards: Exact values can be taken from the accompanying certificate of analysis
Bitte beachten Storage temperature: 2...8 °C. Limited shelf life, expiry date on the label.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9a2cb92efa7aaada832969f3979ca7ab05c9670&vid=158cf7cb9e5969853d02ec084cf71e1b45362187&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b3f90b3fd88177c266e23a460ad3bcc53fbe595&vid=3b3f90b3fd88177c266e23a460ad3bcc53fbe595&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d524253b741051d55e1b2b60d57d5f5423be677f&vid=d524253b741051d55e1b2b60d57d5f5423be677f&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c69b421911954520cc8cfa973e108d2a029d1abd&vid=cbb202a3eb03e8d377861aac0cca5104e68bc5a7&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cac2ffcd8bd8fb1993c9b81a8e9b52fd08232aec&vid=186a9b08459f9a3b6d1bb0ce96569ca1ea505e5e&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c61169229c3f8bb1c1512acfe7a786c3e0628685&vid=7a3fce3763a542dffa7fb4fe3d717dfe1c3134a3&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e3b4814d73c5cb55cf8bacc62744f3ca57436b66&vid=7c9dd24b62384d6860c6939e30553d7c2f4afa7b&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7482f0cb6b98ddc011cfdcd7805c4e68bad0a02&vid=c7482f0cb6b98ddc011cfdcd7805c4e68bad0a02&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e84c466543ded0c430d037ee1189c180ca0fa8aa&vid=e84c466543ded0c430d037ee1189c180ca0fa8aa&prid=web&clid=SAPDAM