Sealing tape, PTFE, Fluo-Kem®, SP Bel-Art
Lieferant: Bel-Art Products, a Part of SP
F240200000
817-1570EA 59.3 EUR
817-1570
Sealing tape, PTFE, Fluo-Kem®, SP Bel-Art
Klebebänder
Non-sintered PTFE tape for use with desiccators, bell jars, glass joints and freeze-drying equipment for tight seal that will not seize over time.
  • Lubricates and seals irregular surfaces
  • Threads and joints without grease or adhesives
Length: 7,6 m
Colour: White
Verpackung: Three per bag.