Zinn(II)chlorid Dihydrat 98,0-103%, Kristalle, AR® ACS, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Gefahr

Synonyme: Zinnsalz

8176-20EA 0 EUR
8176-20
Zinn(II)chlorid Dihydrat 98,0-103%, Kristalle, AR® ACS, Macron Fine Chemicals™
Zinn(II)chlorid Dihydrat
Formel: SnCl₂·2H₂O
Molecular Weight: 225,65 g/mol
Siedepunkt: 623 °C (1013 hPa)
Schmelzpunkt: 246 °C
Dichte: 2,71 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149863
CAS-Nummer: 10025-69-1
EINECS: 231-868-0
UN: 3260
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use 98.0 - 103.0 %
ACS - Assay (SnCl₂·2H₂O) 98.0 - 103.0 %
ACS - Mercury (Hg) ≤0.050 ppm
ACS - Solubility in HCl Passes Test
ACS - Sodium (Na) ≤0.01 %
ACS - Potassium (K) ≤0.005 %
ACS - Iron (Fe) ≤0.003 %
ACS - Calcium (Ca) ≤0.005 %
ACS - Lead (Pb) ≤0.01 %
Appearance: colorless to slightly yellow (but definitely not green) crystals

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR