Notizbücher, Polypaper, Nalgene®
Lieferant: Thermo Fisher Scientific
6300-1000
818-1752EA 102 EUR
818-1752
Notizbücher, Polypaper, Nalgene®
Notizbücher
Laboratory notebooks with uncoated Poly Paper pages and PE cover.
  • Completely waterproof and extremely chemical resistant
  • Pages include spaces for dates, signatures, other pages include instructions for keeping records and table of contents
Format: A4 (24,1×29,2 cm) with 100 numbered pages (50 sheets) printed with a fine green line.