Gefahrenwarnband, 766
Lieferant: 3M
766SG5033
819-0123EA 434 EUR
819-0123
Gefahrenwarnband, 766
Klebebänder
These 766 barrier tapes are made of vinyl with a rubber adhesive and are ideal for hazard warning.
  • Conformable backing ideal for use on curved and irregular surfaces
  • Maintains vivid colour over time for long-lasting quality
  • Rubber adhesive provides good instant adhesion on many surfaces
Used for marking of equipment pinch points or gears for warning purposes, temporary floor marking around equipment or spills.
Länge: 33,0 m