n-Hexan, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Gefahr

8205.1000EA 280 EUR
8205.1000
n-Hexan, BAKER ANALYZED® ACS, J.T.Baker®
n-Hexan
Formel: H₃C(CH₂)₄CH₃
Siedepunkt: 69 °C (1013 hPa)
Schmelzpunkt: –94,3 °C
Dichte: 0,659 g/cm³ (20 °C)
Flammpunkt: –22 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009520
CAS-Nummer: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Exceeds ACS Specifications
Assay (n-hexane) min. 99 %
Assay (sum of 5 isomers, total hexanes plus methylcyclopentane) min. 99 %
Aromatic Compounds (as C6H6) max. 0.02 %
Color (APHA) max. 10
Residue after Evaporation max. 5 ppm
Sulfur Compounds (as S) max. 0.005 %
Thiophene passes test
Water Soluble Titrable Acid (meq/g) max. 0.0002
Ultraviolet Absorbance,(1.00-cm path vs water; curve smooth throughout stated range with no extraneous impurity peaks).
at 210 nm max. 1.00
at 220 nm max. 0.20
at 230 nm max. 0.10
at 240 nm max. 0.04
at 250 nm max. 0.02
at 280-400 nm max. 0.01


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR