ortho-Wolframsäure
Lieferant: Thermo Fisher Scientific
|
Achtung
|
82118.22
82118.14
82118.36
82118.22EA
74.9
EUR
82118.22
82118.14
82118.36
ortho-Wolframsäure
Tungstic(VI) acid
CAS No.: 7783-03-1
Molecular Formula: H2WO4
Formula Weight: 249.86
Physical Form: Powder
Density: 5.5
Refractive Index: 2.24
Melting Point: 100° dec.
Boiling Point: 1473°
Merck Reference: 14,9818
MDL No.: MFCD00011336
Formel:
H₂O₄W Siedepunkt: 1473 °C Dichte: 5,5 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011336 CAS-Nummer: 7783-03-1 EINECS: 231-975-2 Merck Index: 14,9818 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...