Pyronin G, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: Pyronin Y, 3,6-Bis(dimethylamino)xanthyliumchlorid

83200-5G
83200-5GEA 173 EUR
83200-5G
Pyronin G, Sigma-Aldrich®
Pyronin G
Formel: C₁₇H₁₉ClN₂O
Molecular Weight: 302,8 g/mol
Schmelzpunkt: 250…260 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011725
CAS-Nummer: 92-32-0
EINECS: 202-147-8
Merck Index: 12,08190
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37aa2f6c8e135eeb1d582e5d68765c334c759a35&vid=37aa2f6c8e135eeb1d582e5d68765c334c759a35&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR