L(+)-Weinsäure 99,5-101,0%, Pulver Ph. Eur.

Lieferant: VWR Chemicals
Gefahr

83511.290EA 163 EUR
83511.290 83511.360 83511.460
L(+)-Weinsäure 99,5-101,0%, Pulver Ph. Eur.
L(+)-Weinsäure
Formel: C₄H₆O₆
Siedepunkt: 399 °C (1013 hPa)
Schmelzpunkt: 168…170 °C
Dichte: 1,759 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064207
CAS-Nummer: 87-69-4
EINECS: 201-766-0
REACH: 01-2119537204-47
Merck Index: 13,09158
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4e859e52081482a351d52322717614aad450742&vid=879c292903ddee7a59d5d6b2d969c4c8fc6eac75&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (calculated on dried substance) 99.5 - 101.0 %
Appearance Conforms (see CoA/CoS)
Identification A Passes test
Identification B Passes test
Appearance of solution Passes test
Spec. opt. rotation (dried substance) 12.0 - 12.8 °
Oxalic acid Max. 350 ppm
Cl (Chloride) Max. 100 ppm
SO4 (Sulphate) Max. 150 ppm
Ca (Calcium) Max. 200 ppm
Heavy metals (as Pb) Max. 10 ppm
Loss on drying (105°C) Max. 0.2 %
Sulphated ash Max. 0.1 %
Residual solvents Unlikely by manuf.process


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR