Silbernitrat 99-100,5% Ph. Eur.

Lieferant: VWR Chemicals
Gefahr

83517.180EA 718 EUR
83517.180
Silbernitrat 99-100,5% Ph. Eur.
Silbernitrat
Formel: AgNO₃
Siedepunkt: 444 °C (1013 hPa)
Schmelzpunkt: 210 °C
Dichte: 4,35 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003414
CAS-Nummer: 7761-88-8
EINECS: 231-853-9
UN: 1493
ADR: 5.1,II
REACH: 01-2119513705-43
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay 99 - 100.5 %
Appearance Colourless crystals
Identification A Passes test
Identification B Passes test
Appearance of solution Passes test
Acidity or alkalinity Passes test
Foreign salts Max. 0.3 %
Aluminium, lead, copper and bismuth Passes test
Residual solvents Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR