Bismut(III)carbonat basisch Ph. Eur.
Lieferant: VWR Chemicals
Synonyme:
Dibismutcarbonatdioxid, Bismutsubcarbonat, Bismutcarbonatoxid
83518.230EA
117
EUR
83518.230
Bismut(III)carbonat basisch Ph. Eur.
Bismut(III)carbonat basisch
Bitte beachten Not for use as Active Pharmaceutical Ingredient (API)
Formel:
(BiO)₂CO₃ Molecular Weight: 509,97 g/mol Dichte: 6,86 g/cm³ (25 °C) |
MDL:
MFCD00010877 CAS-Nummer: 5892-10-4 EINECS: 227-567-9 Merck Index: 13,01283 |
Spezifikation Testergebnisse
Assay (Bi) (calculated on dried) | 80.0 - 82.5 % |
Appearance | White/almost white powder |
Identification (CO₃) | Passes test |
Identification (Bi) | Passes test |
Appearance of solution | Passes test |
Cl (Chloride) | 500 ppm |
NO₃ (Nitrate) | 0.4 % |
Alkali and alkaline-earth metals | 1.0 % |
Loss on drying (105°C) | 1.0 % |
Residual solvents | Unlikely by manuf.process |
Residual metal catalysts (skip lot): | . |
As (Arsenic) | 5 ppm |
Cd (Cadmium) | 0.15 ppm |
Co (Cobalt) | 1.5 ppm |
Hg (Mercury) | 2.4 ppm |
Ni (Nickel) | 6 ppm |
Pb (Lead) | 20 ppm |
V (Vanadium) | 10 ppm |
Conforms to Ph.Eur. | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...