Thymol Ph. Eur.
Lieferant: VWR Chemicals
|
Gefahr
|
83558.180EA
159
EUR
83558.180
Thymol Ph. Eur.
Thymol
Formel:
2-[(CH₃)₂CH]C₆H₃-5-(CH₃)OH Siedepunkt: 233 °C (1013 hPa) Schmelzpunkt: 49…51 °C Dichte: 0,965 g/cm³ (25 °C) Flammpunkt: 104 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002309 CAS-Nummer: 89-83-8 EINECS: 201-944-8 UN: 2430 ADR: 8,III Merck Index: 13,09476 |
|
Spezifikation Testergebnisse
Appearance | Colourless crystals |
Identification B | Passes test |
Appearance of solution | Passes test |
Acidity | Passes test |
Related substances | Passes test |
Residue on evaporation | Max. 0.05 % |
Residual solvents | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...