D(+)-Saccharose, crystals, AR® ACS, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials
Achtung

Synonyme: Haushaltszucker, D(+)-Sucrose, Rübenzucker, Rohrzucker, 2-β-D-Fructofuranosyl-1-α-D-glucopyranosid, Sucrose, Kristallzucker

8360-24EA 1186.7 EUR
8360-24 8360-20 8360-06
D(+)-Saccharose, crystals, AR® ACS, Macron Fine Chemicals™
D(+)-Saccharose

Meets ACS Reagent Chemical Requirements. For Laboratory, Research, or Manufacturing Use

Formel: C₁₂H₂₂O₁₁
Molecular Weight: 342.3 g/mol
Schmelzpunkt: 169…170 °C
Dichte: 1.29 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00006626
CAS-Nummer: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use Passes Test
Solubility Passes Test
ACS - Insoluble Matter ≤0.005 %
ACS - Chloride (Cl) ≤0.005 %
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Iron (Fe) ≤5 ppm
ACS - Titrable Acid (meq/g) ≤0.0008
ACS - Invert Sugar ≤0.05 %
ACS - Loss on Drying at 105°C ≤0.03 %
ACS - Residue after Ignition ≤0.01 %
ACS - Sulfate and Sulfite (as SO₄) ≤0.005 %
ACS - Specific Rotation [Å]²⁵^D (+) 66.3 - 66.8 Degree


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR