Wasser, HiPerSolv CHROMANORM® für UHPLC-MS
Lieferant: VWR Chemicals
Durch einen 0,1 Micron-Filter gefiltert
83646.290EA
60
EUR
83646.290
Wasser, HiPerSolv CHROMANORM® für UHPLC-MS
Wasser
Formel:
H₂O Siedepunkt: 100 °C (1013 hPa) Schmelzpunkt: 0 °C Dichte: 1 g/cm³ (4 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011332 CAS-Nummer: 7732-18-5 EINECS: 231-791-2 Merck Index: 13,10098 |
Spezifikation Testergebnisse
Clear colorless liquid | complying |
Color (APHA Pt-Co) | max. 5 |
Residue after evaporation | max. 0.7 ppm |
Total organic carbon | max. 10 ppb |
Aluminium (Al) | max. 5 ppb |
Calcium (Ca) | max. 5 ppb |
Iron (Fe) | max. 5 ppb |
Potassium (K) | max. 5 ppb |
Magnesium (Mg) | max. 10 ppb |
Sodium (Na) | max. 25 ppb |
Chloride (Cl) | max. 10 ppb |
Nitrate (NO3) | max. 10 ppb |
Phosphate (PO4) | max. 10 ppb |
Fluorescence 254/450 nm (as Quinine) | max. 0.5 ppb |
Fluorescence 365/450 nm (as Quinine) | max. 0.5 ppb |
UHPLC/MS ESI/APCI+ (as Reserpine) | max. 2 ppb |
UHPLC/MS ESI/APCI- (as Reserpine) | max. 10 ppb |
PEG Signal-to-noise ratio | max. 50 |
UHPLC-UV Gradient (210 nm) | max. 2.0 mAU |
UHPLC-UV Gradient (254 nm) | max. 0.5 mAU |
UV Absorbance 210nm (vs air) | max. 0.01 AU |
UV Absorbance 220 nm (vs air) | max. 0.01 AU |
UV Absorbance 230 nm (vs air) | max. 0.01 AU |
UV Absorbance 240 nm (vs air) | max. 0.01 AU |
UV Absorbance 254 nm (vs air) | max. 0.005 AU |
UV Absorbance 260 nm (vs air) | max. 0.005 AU |
UV Absorbance 280 nm (vs air) | max. 0.005 AU |
Filtered through 0.1 µm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...