Aceton, wasserfrei (max. 0,01% H₂O) ≥99,8%

Lieferant: VWR Chemicals
Gefahr


Filtriert über 0,2 µm-Filter. Unter Stickstoff abgefüllt.

83683.230EA 30.3 EUR
83683.230 83683.290 83683.269
Aceton, wasserfrei (max. 0,01% H₂O) ≥99,8%
Aceton

250 ml bottle with a septum cap featuring six separate re-sealable puncture points


250 and 500 ml bottles with a septum cap featuring six separate re-sealable puncture points

Formel: CH₃COCH₃
Siedepunkt: 56,2 °C (1013 hPa)
Dichte: 0,792 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008765
CAS-Nummer: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3,II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR