Rhodanin, Supelco®

Lieferant: Merck

Synonyme: 2-Thioxo-4-thiazolidinone, 2-Thioxo-1,3-thiazolidin-4-one, 2-Thioxothiazolidin-4-one

M-Clarity™ Qualitätsstufe = MQ100

83700-25G
83700-25GEA 63 EUR
83700-25G
Rhodanin, Supelco®
Rhodanin
Formel: C₃H₃NOS₂
Molecular Weight: 133,19 g/mol
MDL: MFCD00005488
CAS-Nummer: 141-84-4
EINECS: 205-505-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=123abb2ecadbfee04f947d5ad3a2bbfda8483fb4&vid=840be9c7aeab00f464b0db9c9fd7e80d9b140afe&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR