Ammoniaklösung 20%, NORMATOM® Trace_metal

Lieferant: VWR Chemicals
Gefahr

Synonyme: Ammoniumhydroxid, Ammoniak

83870.270EA 105 EUR
83870.270
Ammoniaklösung 20%, NORMATOM® Trace_metal
Ammoniaklösung
Formel: NH₃
Molecular Weight: 35.05 g/mol
Dichte: 0.923 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00066650
CAS-Nummer: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8 III

Bestellen

Spezifikation Testergebnisse

Gehalt 20 - 22 %
Färbung Max. 10 APHA
Cl (Chlorid) Max. 0,5 ppm
PO4 (Phosphat) Max. 0,01 ppm
SO4 (Sulfat) Max. 1 ppm
Ag (Silber) Max. 0,5 ppb
Al (Aluminium) Max. 1 ppb
As (Arsen) Max. 1 ppb
Au (Gold) Max. 0,5 ppb
Ba (Barium) Max. 0,1 ppb
Be (Beryllium) Max. 0,1 ppb
Bi (Bismut) Max. 0,1 ppb
Ca (Calcium) Max. 1 ppb
Cd (Cadmium) Max. 0,5 ppb
Ce (Cer) Max. 0,1 ppb
Co (Cobalt) Max. 0,5 ppb
Cr (Chrom) Max. 0,5 ppb
Cs (Cäsium) Max. 0,1 ppb
Cu (Kupfer) Max. 0,5 ppb
Dy (Dysprosium) Max. 0,1 ppb
Er (Erbium) Max. 0,1 ppb
Eu (Europium) Max. 0,1 ppb
Fe (Eisen) Max. 1 ppb
Ga (Gallium) Max. 0,1 ppb
Gd (Gadolinium) Max. 0,1 ppb
Ge (Germanium) Max. 0,1 ppb
Hg (Quecksilber) Max. 0,2 ppb
Ho (Holmium) Max. 0,1 ppb
In (Indium) Max. 0,1 ppb
K (Kalium) Max. 1 ppb
La (Lanthan) Max. 0,1 ppb
Li (Lithium) Max. 0,1 ppb
Lu (Lutetium) Max. 0,1 ppb
Mg (Magnesium) Max. 1 ppb
Mn (Mangan) Max. 0,5 ppb
Mo (Molybdän) Max. 0,5 ppb
Na (Natrium) Max. 1 ppb
Nb (Niob) Max. 0,1 ppb
Nd (Neodym) Max. 0,1 ppb
Ni (Nickel) Max. 0,5 ppb
Pb (Blei) Max. 0,1 ppb
Pr (Praseodym) Max. 0,1 ppb
Rb (Rubidium) Max. 0,1 ppb
Rh (Rhodium) Max. 0,5 ppb
Sb (Antimon) Max. 0,5 ppb
Sc (Scandium) Max. 0,1 ppb
Se (Selen) Max. 1 ppb
Sm (Samarium) Max. 0,1 ppb
Sn (Zinn) Max. 0,5 ppb
Sr (Strontium) Max. 0,1 ppb
Tb (Terbium) Max. 0,1 ppb
Te (Tellur) Max. 0,1 ppb
Th (Thorium) Max. 0,1 ppb
Ti (Titan) Max. 0,5 ppb
Tl (Thallium) Max. 0,1 ppb
Tm (Thulium) Max. 0,1 ppb
U (Uran) Max. 0,1 ppb
V (Vanadium) Max. 0,5 ppb
W (Wolfram) Max. 0,1 ppb
Y (Yttrium) Max. 0,1 ppb
Yb (Ytterbium) Max. 0,1 ppb
Zn (Zink) Max. 0,5 ppb
Zr (Zirkon) Max. 0,1 ppb


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR