Flußsäure 47%, NORMATOM® für die Spurenanalytik von Metallen
Lieferant: VWR Chemicals
|
Gefahr
|
83873.260EA
163
EUR
83873.260
Flußsäure 47%, NORMATOM® für die Spurenanalytik von Metallen
Hydrofluoric acid
Formel:
HF Molecular Weight: 20,01 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011346 CAS-Nummer: 7664-39-3 EINECS: 231-634-8 UN: 1790 ADR: 8 (6.1),II |
Spezifikation Testergebnisse
Gehalt | 47 - 51 % |
Färbung | Max. 10 APHA |
Cl (Chlorid) | Max. 4 ppm |
SiF6 (Hexafluorosilicat) | Min. 20 ppm |
Gesamt P (Phosphor) | Max. 0,05 ppm |
Gesamt S (Schwefel) | Max. 0,1 ppm |
Ag (Silber) | Max. 0,5 ppb |
Al (Aluminium) | Max. 1 ppb |
As (Arsen) | Max. 0,5 ppb |
Au (Gold) | Max. 0,20 ppb |
B (Bor) | Max. 1 ppb |
Ba (Barium) | Max. 0,1 ppb |
Be (Beryllium) | Max. 0,1 ppb |
Bi (Bismut) | Max. 0,1 ppb |
Ca (Calcium) | Max. 1 ppb |
Cd (Cadmium) | Max. 0,1 ppb |
Ce (Cer) | Max. 0,1 ppb |
Co (Cobalt) | Max. 0,1 ppb |
Cr (Chrom) | Max. 1 ppb |
Cs (Cäsium) | Max. 0,1 ppb |
Cu (Kupfer) | Max. 0,5 ppb |
Dy (Dysprosium) | Max. 0,1 ppb |
Er (Erbium) | Max. 0,1 ppb |
Eu (Europium) | Max. 0,1 ppb |
Fe (Eisen) | Max. 1 ppb |
Ga (Gallium) | Max. 0,1 ppb |
Gd (Gadolinium) | Max. 0,1 ppb |
Ge (Germanium) | Max. 0,1 ppb |
Hf (Hafnium) | Max. 0,1 ppb |
Hg (Quecksilber) | Max. 1 ppb |
Ho (Holmium) | Max. 0,1 ppb |
In (Indium) | Max. 0,1 ppb |
K (Kalium) | Max. 1 ppb |
La (Lanthan) | Max. 0,1 ppb |
Li (Lithium) | Max. 0,1 ppb |
Lu (Lutetium) | Max. 0,1 ppb |
Mg (Magnesium) | Max. 1 ppb |
Mn (Mangan) | Max. 0,1 ppb |
Mo (Molybdän) | Max. 0,1 ppb |
Na (Natrium) | Max. 1 ppb |
Nb (Niob) | Max. 0,1 ppb |
Nd (Neodym) | Max. 0,1 ppb |
Ni (Nickel) | Max. 0,5 ppb |
Pb (Blei) | Max. 0,1 ppb |
Pd (Palladium) | Max. 0,20 ppb |
Pr (Praseodym) | Max. 0,1 ppb |
Pt (Platin) | Max. 0,20 ppb |
Rb (Rubidium) | Max. 0,1 ppb |
Re (Rhenium) | Max. 0,1 ppb |
Rh (Rhodium) | Max. 0,1 ppb |
Ru (Ruthenium) | Max. 0,1 ppb |
Sb (Antimon) | Max. 0,20 ppb |
Sc (Scandium) | Max. 0,1 ppb |
Se (Selen) | Max. 1 ppb |
Sm (Samarium) | Max. 0,1 ppb |
Sn (Zinn) | Max. 0,5 ppb |
Sr (Strontium) | Max. 0,1 ppb |
Tb (Terbium) | Max. 0,1 ppb |
Te (Tellur) | Max. 0,1 ppb |
Th (Thorium) | Max. 0,1 ppb |
Ti (Titan) | Max. 1 ppb |
Tl (Thallium) | Max. 0,1 ppb |
Tm (Thulium) | Max. 0,1 ppb |
U (Uran) | Max. 0,1 ppb |
V (Vanadium) | Max. 0,1 ppb |
W (Wolfram) | Max. 0,5 ppb |
Y (Yttrium) | Max. 0,1 ppb |
Yb (Ytterbium) | Max. 0,1 ppb |
Zn (Zink) | Max. 1 ppb |
Zr (Zirkon) | Max. 0,1 ppb |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...