Perchlorsäure 65%, NORMATOM® für die Spurenanalytik von Metallen

Lieferant: VWR Chemicals
Gefahr

Synonyme: Überchlorsäure

83874.320EA 1130 EUR
83874.320 83874.270
Perchlorsäure 65%, NORMATOM® für die Spurenanalytik von Metallen
Perchlorsäure
Formel: HClO₄
Molecular Weight: 100.46 g/mol
Siedepunkt: 160…181 °C (1013 hPa)
Schmelzpunkt: –112,2 °C
Dichte: 1.53…1.768 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011325
CAS-Nummer: 7601-90-3
EINECS: 231-512-4
UN: 1873
ADR: 5.1 (8) I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fc15d8358229daccc96a56573b32589d2a21575&vid=3780c3029836d8b95181cdef71dc2669f7e2a797&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Gehalt 65 - 71 %
Färbung Max. 10 APHA
Freies Chlor Max. 0,5 ppm
Glührückstand (SO4) Max. 30 ppm
Gesamt N (Stickstoff) Max. 10 ppm
Cl (Chlorid) Max. 3 ppm
ClO3 (Chlorat) Max. 10 ppm
PO4 + SiO4 (als SiO2) Max. 5 ppm
SO4 (Sulfat) Max. 10 ppm
Ag (Silber) Max. 1 ppb
Al (Aluminium) Max. 1 ppb
As (Arsen) Max. 0,5 ppb
Au (Gold) Max. 0,5 ppb
Ba (Barium) Max. 1 ppb
Be (Beryllium) Max. 0,5 ppb
Bi (Bismut) Max. 0,5 ppb
Ca (Calcium) Max. 1 ppb
Cd (Cadmium) Max. 1 ppb
Ce (Cer) Max. 0,5 ppb
Co (Cobalt) Max. 0,5 ppb
Cs (Cäsium) Max. 0,5 ppb
Cu (Kupfer) Max. 0,5 ppb
Dy (Dysprosium) Max. 0,5 ppb
Er (Erbium) Max. 0,5 ppb
Eu (Europium) Max. 0,5 ppb
Fe (Iron) Max. 1 ppb
Ga (Gallium) Max. 0,5 ppb
Gd (Gadolinium) Max. 0,5 ppb
Ho (Holmium) Max. 0,5 ppb
In (Indium) Max. 0,5 ppb
K (Kalium) Max. 1 ppb
La (Lanthan) Max. 0,5 ppb
Li (Lithium) Max. 0,5 ppb
Lu (Lutetium) Max. 0,5 ppb
Mg (Magnesium) Max. 1 ppb
Mn (Mangan) Max. 1 ppb
Mo (Molybdän) Max. 0,5 ppb
Na (Natrium) Max. 1 ppb
Nd (Neodym) Max. 0,5 ppb
Ni (Nickel) Max. 1 ppb
Pb (Blei) Max. 1 ppb
Pd (Palladium) Max. 0,5 ppb
Pr (Praseodym) Max. 0,5 ppb
Pt (Platin) Max. 0,5 ppb
Rb (Rubidium) Max. 0,5 ppb
Rh (Rhodium) Max. 0,5 ppb
Ru (Ruthenium) Max. 0,5 ppb
Sb (Antimon) Max. 0,5 ppb
Sc (Scandium) Max. 0,5 ppb
Sm (Samarium) Max. 0,5 ppb
Sn (Zinn) Max. 1 ppb
Sr (Strontium) Max. 0,5 ppb
Tb (Terbium) Max. 0,5 ppb
Te (Tellur) Max. 0,5 ppb
Th (Thorium) Max. 1 ppb
Ti (Titan) Max. 1 ppb
Tl (Thallium) Max. 0,5 ppb
Tm (Thulium) Max. 0,5 ppb
U (Uran) Max. 0,5 ppb
V (Vanadium) Max. 0,5 ppb
Y (Yttrium) Max. 0,5 ppb
Yb (Ytterbium) Max. 0,5 ppb
Zn (Zink) Max. 1 ppb
Zr (Zirkon) Max. 0,5 ppb


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR