Wasser, NORMATOM® für die Spurenanalytik von Metallen

Lieferant: VWR Chemicals

83877.260EA 156 EUR
83877.260 83877.290
Wasser, NORMATOM® für die Spurenanalytik von Metallen
Wasser
Formel: H₂O
Siedepunkt: 100 °C (1013 hPa)
Schmelzpunkt: 0 °C
Dichte: 1 g/cm³ (4 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Bestellen

Spezifikation Testergebnisse

Färbung 10 CO Max.
Ag (Silber) Max. 50 ppt
Al (Aluminium) Max. 200 ppt
As (Arsen) Max. 50 ppt
Au (Gold) Max. 50 ppt
B (Bor) Max. 500 ppt
Ba (Barium) Max. 50 ppt
Be (Beryllium) Max. 50 ppt
Bi (Bismut) Max. 50 ppt
Ca (Calcium) Max. 500 ppt
Cd (Cadmium) Max. 50 ppt
Ce (Cer) Max. 20 ppt
Co (Cobalt) Max. 50 ppt
Cr (Chrom) Max. 50 ppt
Cs (Cäsium) Max. 50 ppt
Cu (Kupfer) Max. 100 ppt
Dy (Dysprosium) Min. 20 ppt
Er (Erbium) Max. 20 ppt
Eu (Europium) Max. 20 ppt
Fe (Eisen) Max. 300 ppt
Ga (Gallium) Max. 50 ppt
Gd (Gadolinium) Max. 20 ppt
Ge (Germanium) Max. 50 ppt
Hf (Hafnium) Max. 10 ppt
Hg (Quecksilber) Max. 200 ppt
Ho (Holmium) Max. 20 ppt
In (Indium) Max. 50 ppt
K (Kalium) Max. 300 ppt
La (Lanthan) Max. 20 ppt
Li (Lithium) Max. 50 ppt
Lu (Lutetium) Max. 20 ppt
Mg (Magnesium) Max. 100 ppt
Mn (Mangan) Max. 50 ppt
Mo (Molybdän) Max. 50 ppt
Na (Natrium) Max. 500 ppt
Nb (Niob) Max. 20 ppt
Nd (Neodym) Max. 20 ppt
Ni (Nickel) Max. 50 ppt
Pb (Blei) Max. 50 ppt
Pd (Palladium) Max. 50 ppt
Pr (Praseodym) Max. 20 ppt
Pt (Platin) Max. 50 ppt
Rb (Rubidium) Max. 20 ppt
Re (Rhenium) Max. 50 ppt
Rh (Rhodium) Max. 50 ppt
Ru (Ruthenium) Max. 50 ppt
Sb (Antimon) Max. 50 ppt
Sc (Scandium) Max. 20 ppt
Se (Selen) Max. 200 ppt
Sm (Samarium) Max. 20 ppt
Sn (Zinn) Max. 50 ppt
Sr (Strontium) Max. 50 ppt
Ta (Tantal) Max. 50 ppt
Tb (Terbium) Max. 20 ppt
Te (Tellur) Max. 50 ppt
Th (Thorium) Max. 10 ppt
Ti (Titan) Max. 50 ppt
Tl (Thallium) Max. 50 ppt
Tm (Thulium) Max. 20 ppt
U (Uran) Max. 10 ppt
V (Vanadium) Max. 50 ppt
W (Wolfram) Max. 100 ppt
Y (Yttrium) Max. 10 ppt
Yb (Ytterbium) Max. 20 ppt
Zn (Zink) Max. 500 ppt
Zr (Zirkon) Max. 50 ppt


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR