Ruthenium standard for AAS, analytical standard, Supelco®
Lieferant: Merck
84033-100ML
84033-100MLEA 39 EUR
84033-100ML
Ruthenium standard for AAS, analytical standard, Supelco®
Standards Kationischer Standards Ruthenium-Standards
Ruthenium standard solution may be used in the determination of ruthenium ions in uranium alloys using atomic absorption spectrophotometry.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ae6666921216796b31d4bb82dcc898a3bcbfc7f&vid=2ae6666921216796b31d4bb82dcc898a3bcbfc7f&prid=web&clid=SAPDAM